Gene Transcripts

Gene Symbol
OR8S1
Entrez Gene ID
341568
Gene Name
olfactory receptor family 8 subfamily S member 1
Gene Aliases
Chromosome Location
Chr. 12: 48525632 - 48528103 on Build GRCh38
UniGene ID
Hs.524373
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005203.2 NP_001005203.2 1 409

View Full Product Details