Gene Transcripts
- Gene Symbol
- GLDN
- Entrez Gene ID
- 342035
- Gene Name
- gliomedin
- Gene Aliases
- CLOM, COLM, CRG-L2, CRGL2, UNC-112
- Chromosome Location
- Chr. 15: 51341516 - 51413365 on Build GRCh38
- UniGene ID
- Hs.526441
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_181789.2 |
NP_861454.2
|
8, 9 |
1202 |
|
XM_011521501.2 |
XP_011519803.1
|
8, 9 |
3059 |
|
XM_017022121.1 |
XP_016877610.1
|
7, 8 |
1045 |
|
XM_017022122.1 |
XP_016877611.1
|
8, 9 |
1233 |
|
XM_017022123.1 |
XP_016877612.1
|
8, 9 |
1024 |
|
XM_017022124.1 |
XP_016877613.1
|
8, 9 |
1076 |
|
XM_017022125.1 |
XP_016877614.1
|
8, 9 |
1063 |
|
XM_017022126.1 |
XP_016877615.1
|
8, 9 |
1076 |
|
GenBank mRNA |
AK172756.1 |
- |
8, 9 |
1038 |
|
AY358144.1 |
- |
7, 8 |
663 |
|
BC113397.1 |
- |
8, 9 |
688 |
8322449 |
BC113399.1 |
- |
8, 9 |
688 |
8322451 |
BX538105.1 |
- |
8, 9 |
711 |
|
View Full Product Details