Gene Transcripts

Gene Symbol
SMTNL2
Entrez Gene ID
342527
Gene Name
smoothelin like 2
Gene Aliases
Chromosome Location
Chr. 17: 4583981 - 4608319 on Build GRCh38
UniGene ID
Hs.441709
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114974.1 NP_001108446.1 8 1769
NM_198501.2 NP_940903.2 8 1421
XM_005256622.3 XP_005256679.1 7 1598
XM_006721515.2 XP_006721578.1 8 1937
GenBank mRNA AK124452.1 - 8 1403
BC110859.1 - 8 1769 5190568

View Full Product Details