Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145028.1 | NP_001138500.1 | 3 | 260 | |
XM_005259891.4 | XP_005259948.1 | 4 | 461 | ||
XM_011527965.2 | XP_011526267.1 | 3 | 381 | ||
GenBank mRNA | AW130597.1 | - | 1 | 238 | |
CR747190.1 | - | 2 | 383 |