Gene Transcripts
- Gene Symbol
- GPATCH11
- Entrez Gene ID
- 253635
- Gene Name
- G-patch domain containing 11
- Gene Aliases
- CCDC75, CENP-Y, CENPY
- Chromosome Location
- Chr. 2: 37084451 - 37099244 on Build GRCh38
- UniGene ID
- Hs.595250
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278505.1 |
NP_001265434.1
|
6, 7 |
593 |
|
NM_001322249.1 |
NP_001309178.1
|
7, 8 |
820 |
|
NM_174931.3 |
NP_777591.3
|
8, 9 |
862 |
|
XM_017003748.1 |
XP_016859237.1
|
8, 9 |
763 |
|
GenBank mRNA |
AK095667.1 |
- |
6, 7 |
526 |
|
AK289504.1 |
- |
6, 7 |
508 |
|
AK294697.1 |
- |
8, 9 |
776 |
|
BC071798.1 |
- |
6, 7 |
525 |
|
BM285380.1 |
- |
8, 9 |
862 |
|
BX647651.1 |
- |
2, 3 |
154 |
|
View Full Product Details