Gene Transcripts

Gene Symbol
OAS1
Entrez Gene ID
4938
Gene Name
2'-5'-oligoadenylate synthetase 1
Gene Aliases
E18/E16, IFI-4, OIAS, OIASI
Chromosome Location
Chr. 12: 112906777 - 112932190 on Build GRCh38
UniGene ID
Hs.524760
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001032409.2 NP_001027581.1 2 586
NM_001320151.1 NP_001307080.1 2 586
NM_002534.3 NP_002525.2 2 586
NM_016816.3 NP_058132.2 2 586
XM_006719434.1 XP_006719497.1 2 413
XM_011538413.1 XP_011536715.1 2 413
XM_017019361.1 XP_016874850.1 2 413
XM_017019362.1 XP_016874851.1 2 285
GenBank mRNA AJ629455.1 - 2 356
AK223006.1 - 2 345
AK225384.1 - 2 386
AK291003.1 - 2 390
AK300346.1 - 2 311
AK301498.1 - 2 345
AY730627.1 - 2 332
AY730628.1 - 2 332
BC000562.2 - 2 393 3162421
BC061587.1 - 2 354 6502031
BT006785.1 - 2 323
D00068.1 - 2 386
X02874.1 - 2 356
X04371.1 - 2 416

View Full Product Details