Gene Transcripts

Gene Symbol
IGFBPL1
Entrez Gene ID
347252
Gene Name
insulin like growth factor binding protein like 1
Gene Aliases
IGFBP-RP4, IGFBPRP4, bA113O24.1
Chromosome Location
Chr. 9: 38406528 - 38424447 on Build GRCh38
UniGene ID
Hs.349705
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007563.2 NP_001007564.1 4 798
GenBank mRNA AK131521.1 - 4 351

View Full Product Details