Gene Transcripts
- Gene Symbol
- ITIH6
- Entrez Gene ID
- 347365
- Gene Name
- inter-alpha-trypsin inhibitor heavy chain family member 6
- Gene Aliases
- ITIH5L, UNQ6369, dJ14O9.1
- Chromosome Location
- Chr. X: 54748899 - 54798240 on Build GRCh38
- UniGene ID
- Hs.454272
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_198510.2 |
NP_940912.1
|
12 |
3512 |
|
GenBank mRNA |
AY358170.1 |
- |
12 |
3512 |
|
View Full Product Details