Gene Transcripts
- Gene Symbol
- CENPVP1
- Entrez Gene ID
- 389857
- Gene Name
- centromere protein V pseudogene 1
- Gene Aliases
- CENPVL1, PRR6L1
- Chromosome Location
- Chr. X: 51710829 - 51712131 on Build GRCh38
- UniGene ID
- Hs.730388
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033772.1 |
|
1 |
239 |
|
GenBank mRNA |
AY730278.1 |
- |
1 |
239 |
|
- Gene Symbol
- CENPVP2
- Entrez Gene ID
- 441495
- Gene Name
- centromere protein V pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. X: 51681212 - 51682514 on Build GRCh38
- UniGene ID
- Hs.187774
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033773.1 |
|
1 |
239 |
|
View Full Product Details