Gene Transcripts
- Gene Symbol
- FTCDNL1
- Entrez Gene ID
- 348751
- Gene Name
- formiminotransferase cyclodeaminase N-terminal like
- Gene Aliases
- FONG
- Chromosome Location
- Chr. 2: 199760536 - 199851173 on Build GRCh38
- UniGene ID
- Hs.734578
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_034096.1 |
|
2 |
514 |
|
GenBank mRNA |
AB568489.1 |
- |
2 |
461 |
|
BC039445.1 |
- |
2 |
514 |
|
View Full Product Details