Gene Transcripts

Gene Symbol
HDDC3
Entrez Gene ID
374659
Gene Name
HD domain containing 3
Gene Aliases
(ppGpp)ase, MESH1
Chromosome Location
Chr. 15: 90930918 - 90932569 on Build GRCh38
UniGene ID
Hs.349979
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286451.1 NP_001273380.1 2 184
NM_198527.3 NP_940929.1 2 184
NR_104447.1 2 227
GenBank mRNA AK057584.1 - 1 495
AW138983.1 - 2 184
BC033794.1 - 2 184 5122164
BQ777217.1 - 2 134
CV573934.1 - 2 169

View Full Product Details