Gene Transcripts

Gene Symbol
PTAR1
Entrez Gene ID
375743
Gene Name
protein prenyltransferase alpha subunit repeat containing 1
Gene Aliases
Chromosome Location
Chr. 9: 69709522 - 69760050 on Build GRCh38
UniGene ID
Hs.744192
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099666.1 NP_001093136.1 2 145
XM_005251976.4 XP_005252033.1 2 235
XM_005251977.4 XP_005252034.1 2 209
XM_005251980.4 XP_005252037.1 2 195
XM_005251981.4 XP_005252038.1 2 195
XM_011518640.1 XP_011516942.1 2 229
GenBank mRNA BC053622.1 - 2 127

View Full Product Details