Gene Transcripts

Gene Symbol
GPR153
Entrez Gene ID
387509
Gene Name
G protein-coupled receptor 153
Gene Aliases
PGR1
Chromosome Location
Chr. 1: 6247346 - 6260975 on Build GRCh38
UniGene ID
Hs.531581
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207370.2 NP_997253.2 4 1174
XM_011541434.2 XP_011539736.1 4 1278
XM_017001250.1 XP_016856739.1 3 1260
XM_017001251.1 XP_016856740.1 4 1163
GenBank mRNA AY255529.1 - 2 437
BC068275.1 - 4 1174 5167285

View Full Product Details