Gene Transcripts
- Gene Symbol
- FAM21A
- Entrez Gene ID
- 387680
- Gene Name
- family with sequence similarity 21 member A
- Gene Aliases
- FAM21B, bA56A21.1, bA98I6.1
- Chromosome Location
- Chr. 10: 50067888 - 50133509 on Build GRCh38
- UniGene ID
- Hs.365286
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001005751.2 |
NP_001005751.1
|
8, 9 |
847 |
|
NM_001291398.1 |
NP_001278327.1
|
8, 9 |
847 |
|
XM_005269806.3 |
XP_005269863.2
|
8, 9 |
795 |
|
XM_005269807.3 |
XP_005269864.2
|
8, 9 |
795 |
|
XM_005269808.3 |
XP_005269865.2
|
8, 9 |
795 |
|
XM_006717831.3 |
XP_006717894.2
|
8, 9 |
795 |
|
XM_006717832.3 |
XP_006717895.2
|
8, 9 |
795 |
|
XM_011539786.2 |
XP_011538088.1
|
8, 9 |
795 |
|
XM_017016227.1 |
XP_016871716.1
|
8, 9 |
795 |
|
XM_017016228.1 |
XP_016871717.1
|
8, 9 |
795 |
|
XM_017016229.1 |
XP_016871718.1
|
8, 9 |
795 |
|
XM_017016230.1 |
XP_016871719.1
|
7, 8 |
660 |
|
XM_017016231.1 |
XP_016871720.1
|
8, 9 |
795 |
|
XM_017016232.1 |
XP_016871721.1
|
8, 9 |
795 |
|
XM_017016233.1 |
XP_016871722.1
|
5, 6 |
370 |
|
XM_017016234.1 |
XP_016871723.1
|
9, 10 |
889 |
|
GenBank mRNA |
AK001686.1 |
- |
5, 6 |
498 |
|
AK295129.1 |
- |
8, 9 |
799 |
|
AK304116.1 |
- |
8, 9 |
783 |
|
AK311628.1 |
- |
5, 6 |
401 |
|
BC075815.1 |
- |
5, 6 |
412 |
|
BC082258.1 |
- |
8, 9 |
774 |
6278433 |
BC144479.1 |
- |
8, 9 |
876 |
9053006 |
DC403380.1 |
- |
5, 6 |
401 |
|
- Gene Symbol
- FAM21C
- Entrez Gene ID
- 253725
- Gene Name
- family with sequence similarity 21 member C
- Gene Aliases
- FAM21A, VPEF
- Chromosome Location
- Chr. 10: 45727170 - 45792964 on Build GRCh38
- UniGene ID
- Hs.365286
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC150611.1 |
- |
8, 9 |
812 |
|
BM126842.1 |
- |
4, 5 |
302 |
|
View Full Product Details