Gene Transcripts

Gene Symbol
SLC9A4
Entrez Gene ID
389015
Gene Name
solute carrier family 9 member A4
Gene Aliases
NHE4
Chromosome Location
Chr. 2: 102473282 - 102533974 on Build GRCh38
UniGene ID
Hs.447686
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011552.3 NP_001011552.2 8 2041
XM_011511158.1 XP_011509460.1 8 1975
GenBank mRNA CR627411.1 - 8 2042

View Full Product Details