Gene Transcripts
- Gene Symbol
- OR52I1
- Entrez Gene ID
- 390037
- Gene Name
- olfactory receptor family 52 subfamily I member 1
- Gene Aliases
- OR11-13
- Chromosome Location
- Chr. 11: 4594039 - 4595013 on Build GRCh38
- UniGene ID
- Hs.553727
- Species
- Homo sapiens
- Gene Symbol
- OR52I2
- Entrez Gene ID
- 143502
- Gene Name
- olfactory receptor family 52 subfamily I member 2
- Gene Aliases
- OR11-12, OR52I1
- Chromosome Location
- Chr. 11: 4586791 - 4587905 on Build GRCh38
- UniGene ID
- Hs.553607
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC136921.1 |
- |
1 |
248 |
|
BC151144.1 |
- |
1 |
248 |
9020913 |
View Full Product Details