Gene Transcripts

Gene Symbol
OR52D1
Entrez Gene ID
390066
Gene Name
olfactory receptor family 52 subfamily D member 1
Gene Aliases
OR11-43
Chromosome Location
Chr. 11: 5488685 - 5489749 on Build GRCh38
UniGene ID
Hs.553734
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005163.2 NP_001005163.1 1 70
GenBank mRNA BC136928.1 - 1 70
BC136930.1 - 1 70 9020922

View Full Product Details