Gene Transcripts

Gene Symbol
OR52E8
Entrez Gene ID
390079
Gene Name
olfactory receptor family 52 subfamily E member 8
Gene Aliases
OR11-54
Chromosome Location
Chr. 11: 5856749 - 5857702 on Build GRCh38
UniGene ID
Hs.553738
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005168.1 NP_001005168.1 1 629
GenBank mRNA BC137030.1 - 1 661
BC137041.1 - 1 661 9021037

View Full Product Details