Gene Transcripts

Gene Symbol
OR5D16
Entrez Gene ID
390144
Gene Name
olfactory receptor family 5 subfamily D member 16
Gene Aliases
OR11-154
Chromosome Location
Chr. 11: 55838752 - 55839738 on Build GRCh38
UniGene ID
Hs.554527
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005496.1 NP_001005496.1 1 328

View Full Product Details