Gene Transcripts

Gene Symbol
OR8H3
Entrez Gene ID
390152
Gene Name
olfactory receptor family 8 subfamily H member 3
Gene Aliases
OR11-172
Chromosome Location
Chr. 11: 56122373 - 56123311 on Build GRCh38
UniGene ID
Hs.553745
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005201.1 NP_001005201.1 1 886

View Full Product Details