Gene Transcripts

Gene Symbol
OR6C76
Entrez Gene ID
390326
Gene Name
olfactory receptor family 6 subfamily C member 76
Gene Aliases
Chromosome Location
Chr. 12: 55426254 - 55427192 on Build GRCh38
UniGene ID
Hs.553764
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005183.1 NP_001005183.1 1 816

View Full Product Details