Gene Transcripts

Gene Symbol
OR3A4P
Entrez Gene ID
390756
Gene Name
olfactory receptor family 3 subfamily A member 4 pseudogene
Gene Aliases
OLFRA05, OLFRA06, OR17-13, OR17-16, OR17-24, OR17-25, OR24, OR25, OR3A4, OR3A5P
Chromosome Location
Chr. 17: 3310245 - 3311446 on Build GRCh38
UniGene ID
Hs.632245
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024128.1 1 543
GenBank mRNA BC136874.1 - 1 543
BC136875.1 - 1 543

View Full Product Details