Gene Transcripts

Gene Symbol
OR2L8
Entrez Gene ID
391190
Gene Name
olfactory receptor family 2 subfamily L member 8 (gene/pseudogene)
Gene Aliases
Chromosome Location
Chr. 1: 247948858 - 247949796 on Build GRCh38
UniGene ID
Hs.690212
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001963.1 NP_001001963.1 1 63
GenBank mRNA BC140751.1 - 1 144

View Full Product Details