Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001460.4 | NP_001451.2 | 5 | 707 | |
GenBank mRNA | AK312877.1 | - | 5 | 613 | |
AL833218.1 | - | 5 | 596 | ||
BC005894.1 | - | 5 | 613 | 4293031 | |
BT006979.1 | - | 4 | 549 | ||
Y09267.1 | - | 5 | 609 |