Gene Transcripts

Gene Symbol
PRAMEF5
Entrez Gene ID
343068
Gene Name
PRAME family member 5
Gene Aliases
PRAMEF23, PRAMEF5L
Chromosome Location
Chr. 1: 13254198 - 13263435 on Build GRCh38
UniGene ID
Hs.567864
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013407.2 NP_001013425.2 4 1328
GenBank mRNA BC101344.1 - 4 1314
BC130274.1 - 4 1313 40146305
BC130276.1 - 4 1313 40146307

Gene Symbol
PRAMEF4
Entrez Gene ID
400735
Gene Name
PRAME family member 4
Gene Aliases
Chromosome Location
Chr. 1: 12879212 - 12886201 on Build GRCh38
UniGene ID
Hs.551617
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009611.4 NP_001009611.2 4 1305
GenBank mRNA AL049681.1 - 4 1305

Gene Symbol
PRAMEF6
Entrez Gene ID
440561
Gene Name
PRAME family member 6
Gene Aliases
Chromosome Location
Chr. 1: 12938472 - 12947580 on Build GRCh38
UniGene ID
Hs.567864
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010889.2 NP_001010889.1 4 1314
GenBank mRNA BC101342.2 - 4 1314

Gene Symbol
PRAMEF9
Entrez Gene ID
343070
Gene Name
PRAME family member 9
Gene Aliases
Chromosome Location
Chr. 1: 13175281 - 13179132 on Build GRCh38
UniGene ID
Hs.631873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK124292.1 - 4 1328

View Full Product Details