Gene Transcripts

Gene Symbol
NBPF8
Entrez Gene ID
728841
Gene Name
neuroblastoma breakpoint family member 8
Gene Aliases
NBPF8P
Chromosome Location
Chr. 1: 120436351 - 120467844 on Build GRCh38
UniGene ID
Hs.445080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037501.2 NP_001032590.2 3 496
NR_102404.1 3 496
NR_102405.1 2 393
GenBank mRNA AY894567.1 - 2 393
AY894568.1 - 3 291
AY894571.1 - 2 393
AY894572.1 - 3 496
AY894573.1 - 2 393
BC086308.1 - 3 339 5453529

Gene Symbol
NBPF10
Entrez Gene ID
100132406
Gene Name
neuroblastoma breakpoint family member 10
Gene Aliases
AB6, AG1, NBPF9
Chromosome Location
Chr. 1: 146066318 - 146144804 on Build GRCh38
UniGene ID
Hs.714127
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BX538005.1 - 9 1494

Gene Symbol
NBPF26
Entrez Gene ID
101060684
Gene Name
neuroblastoma breakpoint family member 26
Gene Aliases
Chromosome Location
Chr. 1: 120805561 - 120842215 on Build GRCh38
UniGene ID
Hs.534675
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011542529.2 XP_011540831.1 3 354
XM_017003011.1 XP_016858500.1 3 353

Gene Symbol
NBPF9
Entrez Gene ID
400818
Gene Name
neuroblastoma breakpoint family member 9
Gene Aliases
AE01
Chromosome Location
Chr. 1: 149054033 - 149090861 on Build GRCh38
UniGene ID
Hs.445080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037675.3 NP_001032764.2 4 399
NM_001277444.1 NP_001264373.1 4 399
XM_006711317.2 XP_006711380.1 5 627
XM_017001278.1 XP_016856767.1 5 627
GenBank mRNA AY894574.1 - 4 401

View Full Product Details