Gene Transcripts

Gene Symbol
C2orf66
Entrez Gene ID
401027
Gene Name
chromosome 2 open reading frame 66
Gene Aliases
IIDS6411, UNQ6411
Chromosome Location
Chr. 2: 196804415 - 196810276 on Build GRCh38
UniGene ID
Hs.245666
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_213608.2 NP_998773.1 3 1323
GenBank mRNA AY358249.1 - 3 1323
BC137150.1 - 3 1199 9021147
BC137151.1 - 3 1199 9021148

View Full Product Details