Gene Transcripts

Gene Symbol
ASB18
Entrez Gene ID
401036
Gene Name
ankyrin repeat and SOCS box containing 18
Gene Aliases
ASB-18
Chromosome Location
Chr. 2: 236194872 - 236264345 on Build GRCh38
UniGene ID
Hs.639909
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212556.2 NP_997721.2 2 281
GenBank mRNA AK123854.1 - 1 289

View Full Product Details