Gene Transcripts

Gene Symbol
TOMM5
Entrez Gene ID
401505
Gene Name
translocase of outer mitochondrial membrane 5
Gene Aliases
C9orf105, Tom5, bA613M10.3
Chromosome Location
Chr. 9: 37588413 - 37592639 on Build GRCh38
UniGene ID
Hs.130774
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001790.2 NP_001001790.1 2 482
NM_001134484.1 NP_001127956.1 2 641
NM_001134485.1 NP_001127957.1 2 584
GenBank mRNA BC034247.1 - 2 414

View Full Product Details