Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001463.3 | NP_001454.2 | 3 | 792 | |
GenBank mRNA | AK130009.1 | - | 4 | 444 | |
AK312741.1 | - | 3 | 671 | ||
BC027855.1 | - | 3 | 644 | 5221358 | |
BT019883.1 | - | 3 | 574 | ||
U24163.1 | - | 3 | 782 | ||
U68057.1 | - | 3 | 634 | ||
U91903.1 | - | 3 | 643 |