Gene Transcripts
- Gene Symbol
- FAM66C
- Entrez Gene ID
- 440078
- Gene Name
- family with sequence similarity 66 member C
- Gene Aliases
- Chromosome Location
- Chr. 12: 8180209 - 8201000 on Build GRCh38
- UniGene ID
- Hs.512257
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026788.1 |
|
7 |
1804 |
|
GenBank mRNA |
BC048994.1 |
- |
5 |
1161 |
|
View Full Product Details