Gene Transcripts

Gene Symbol
DBX2
Entrez Gene ID
440097
Gene Name
developing brain homeobox 2
Gene Aliases
Chromosome Location
Chr. 12: 45014756 - 45051099 on Build GRCh38
UniGene ID
Hs.302764
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004329.2 NP_001004329.2 2, 3 663
GenBank mRNA AK131230.1 - 2, 3 663

View Full Product Details