Gene Transcripts

Gene Symbol
HSBP1L1
Entrez Gene ID
440498
Gene Name
heat shock factor binding protein 1 like 1
Gene Aliases
Chromosome Location
Chr. 18: 79964582 - 79970822 on Build GRCh38
UniGene ID
Hs.191582
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136180.1 NP_001129652.1 2 253
GenBank mRNA BC017654.1 - 2 181
BC040056.2 - 2 182
BC157848.1 - 2 175 9057067
DA919112.1 - 2 253

View Full Product Details