Gene Transcripts

Gene Symbol
TPBGL
Entrez Gene ID
100507050
Gene Name
trophoblast glycoprotein-like
Gene Aliases
Chromosome Location
Chr. 11: 75240905 - 75243704 on Build GRCh38
UniGene ID
Hs.577323
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195528.1 NP_001182457.1 1 1193

View Full Product Details