Gene Transcripts

Gene Symbol
IGFL4
Entrez Gene ID
444882
Gene Name
IGF like family member 4
Gene Aliases
Chromosome Location
Chr. 19: 46039748 - 46041016 on Build GRCh38
UniGene ID
Hs.531310
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002923.1 NP_001002923.1 2 101
GenBank mRNA AY672114.1 - 2 101

View Full Product Details