Gene Transcripts

Gene Symbol
FRG2
Entrez Gene ID
448831
Gene Name
FSHD region gene 2
Gene Aliases
FRG2A
Chromosome Location
Chr. 4: 190024367 - 190027257 on Build GRCh38
UniGene ID
Hs.626907
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005217.1 NP_001005217.1 4 1775
NM_001286820.1 NP_001273749.1 4 1778
GenBank mRNA AY714545.1 - 4 1775

View Full Product Details