Gene Transcripts

Gene Symbol
C8orf22
Entrez Gene ID
492307
Gene Name
chromosome 8 open reading frame 22
Gene Aliases
Chromosome Location
Chr. 8: 49054336 - 49076083 on Build GRCh38
UniGene ID
Hs.49890
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007176.4 NP_001007177.1 2 231
NM_001256596.1 NP_001243525.1 2 123
NM_001256597.1 NP_001243526.1 2 231
NM_001256598.1 NP_001243527.1 2 231
GenBank mRNA AJ276240.1 - 2 218
BC017981.1 - 2 224 4245342
BG202040.1 - 2 149
DA820911.1 - 2 232
DA900243.1 - 2 231

View Full Product Details