Gene Transcripts

Gene Symbol
COA5
Entrez Gene ID
493753
Gene Name
cytochrome c oxidase assembly factor 5
Gene Aliases
6330578E17Rik, C2orf64, CEMCOX3, Pet191
Chromosome Location
Chr. 2: 98599323 - 98608518 on Build GRCh38
UniGene ID
Hs.596537
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008215.2 NP_001008216.1 2, 3 293
GenBank mRNA AK131034.1 - 2, 3 290
BC047722.1 - 2, 3 267 5748383
DA214157.1 - 2, 3 295

View Full Product Details