Gene Transcripts

Gene Symbol
YIF1B
Entrez Gene ID
90522
Gene Name
Yip1 interacting factor homolog B, membrane trafficking protein
Gene Aliases
FinGER8
Chromosome Location
Chr. 19: 38303560 - 38315976 on Build GRCh38
UniGene ID
Hs.280741
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039671.2 NP_001034760.1 8 1826
NM_001039672.2 NP_001034761.1 8 1844
NM_001039673.2 NP_001034762.1 8 1835
NM_001145461.1 NP_001138933.1 8 1820
NM_001145462.1 NP_001138934.1 8 1796
NM_001145463.1 NP_001138935.1 9 1912
GenBank mRNA AK027130.1 - 1 519
BC025257.1 - 1 455

View Full Product Details