Gene Transcripts

Gene Symbol
HEPN1
Entrez Gene ID
641654
Gene Name
hepatocellular carcinoma, down-regulated 1
Gene Aliases
Chromosome Location
Chr. 11: 124919250 - 124920677 on Build GRCh38
UniGene ID
Hs.159863
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037558.2 NP_001032647.2 1 1248
GenBank mRNA AY275431.1 - 1 1244

View Full Product Details