Gene Transcripts
- Gene Symbol
- GOLGA6L7P
- Entrez Gene ID
- 728310
- Gene Name
- golgin A6 family-like 7, pseudogene
- Gene Aliases
- GOLGA6L7
- Chromosome Location
- Chr. 15: 28844961 - 28848675 on Build GRCh38
- UniGene ID
- Hs.589351
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_047567.1 |
|
2 |
227 |
|
GenBank mRNA |
AK302238.1 |
- |
2 |
227 |
|
View Full Product Details