Gene Transcripts

Gene Symbol
KRTAP27-1
Entrez Gene ID
643812
Gene Name
keratin associated protein 27-1
Gene Aliases
Chromosome Location
Chr. 21: 30337013 - 30337694 on Build GRCh38
UniGene ID
Hs.580881
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077711.1 NP_001071179.1 1 236
GenBank mRNA AB096937.1 - 1 236

View Full Product Details