Gene Transcripts
- Gene Symbol
- PGA4
- Entrez Gene ID
- 643847
- Gene Name
- pepsinogen 4, group I (pepsinogen A)
- Gene Aliases
- Chromosome Location
- Chr. 11: 61222349 - 61231695 on Build GRCh38
- UniGene ID
- Hs.661883
- Species
- Homo sapiens
- Gene Symbol
- ARMC8
- Entrez Gene ID
- 25852
- Gene Name
- armadillo repeat containing 8
- Gene Aliases
- GID5, HSPC056, S863-2, VID28
- Chromosome Location
- Chr. 3: 138187248 - 138298389 on Build GRCh38
- UniGene ID
- Hs.744868
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098500.1 |
- |
|
3126 |
|
- Gene Symbol
- PGA3
- Entrez Gene ID
- 643834
- Gene Name
- pepsinogen 3, group I (pepsinogen A)
- Gene Aliases
- Chromosome Location
- Chr. 11: 61203513 - 61212869 on Build GRCh38
- UniGene ID
- Hs.601055
- Species
- Homo sapiens
- Gene Symbol
- PGA5
- Entrez Gene ID
- 5222
- Gene Name
- pepsinogen 5, group I (pepsinogen A)
- Gene Aliases
- Pg5
- Chromosome Location
- Chr. 11: 61241176 - 61251444 on Build GRCh38
- UniGene ID
- Hs.432854
- Species
- Homo sapiens
View Full Product Details