Gene Transcripts

Gene Symbol
PGA3
Entrez Gene ID
643834
Gene Name
pepsinogen 3, group I (pepsinogen A)
Gene Aliases
Chromosome Location
Chr. 11: 61203513 - 61212869 on Build GRCh38
UniGene ID
Hs.601055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079807.3 NP_001073275.1 8 1028
GenBank mRNA AK125628.1 - 8 1042
AK225669.1 - 8 1026
AK225674.1 - 8 1054
AK225679.1 - 8 1026
AK301286.1 - 5 503
AK301293.1 - 6 629
AK312941.1 - 8 1028
AL832946.1 - 1 1431
BC171815.1 - 8 1022 9054469

Gene Symbol
PGA4
Entrez Gene ID
643847
Gene Name
pepsinogen 4, group I (pepsinogen A)
Gene Aliases
Chromosome Location
Chr. 11: 61222349 - 61231695 on Build GRCh38
UniGene ID
Hs.661883
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079808.3 NP_001073276.1 8 1028
GenBank mRNA AK291864.1 - 8 1026
AK301305.1 - 5 500
BC150659.1 - 8 1022 9057030
BC171808.1 - 8 1022 9054462
BC171814.1 - 8 1022 9054468
BC171910.1 - 8 1022 9054564
BC171920.1 - 8 1022 9054574

Gene Symbol
PGA5
Entrez Gene ID
5222
Gene Name
pepsinogen 5, group I (pepsinogen A)
Gene Aliases
Pg5
Chromosome Location
Chr. 11: 61241176 - 61251444 on Build GRCh38
UniGene ID
Hs.432854
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014224.4 NP_055039.1 8 1027
GenBank mRNA BC029055.1 - 8 1006
BC146999.1 - 8 1027 9056927
BC171889.1 - 8 1027 9054543
BC171897.1 - 8 1027 9054551

View Full Product Details