Gene Transcripts
- Gene Symbol
- MEIG1
- Entrez Gene ID
- 644890
- Gene Name
- meiosis/spermiogenesis associated 1
- Gene Aliases
- SPATA39, bA2K17.3
- Chromosome Location
- Chr. 10: 14959439 - 14972851 on Build GRCh38
- UniGene ID
- Hs.257249
- Species
- Homo sapiens
- Gene Symbol
- LOC105376430
- Entrez Gene ID
- 105376430
- Gene Name
- uncharacterized LOC105376430
- Gene Aliases
- Chromosome Location
- Chr. 10: 14972564 - 14988050 on Build GRCh38
- UniGene ID
- Hs.615546
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_134493.1 |
|
1 |
14 |
|
GenBank mRNA |
BG189096.1 |
- |
1 |
33 |
|
View Full Product Details