Gene Transcripts
- Gene Symbol
- EIF3D
- Entrez Gene ID
- 8664
- Gene Name
- eukaryotic translation initiation factor 3 subunit D
- Gene Aliases
- EIF3S7, eIF3-p66, eIF3-zeta
- Chromosome Location
- Chr. 22: 36510850 - 36529431 on Build GRCh38
- UniGene ID
- Hs.55682
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BG718662.1 |
- |
1 |
141 |
|
View Full Product Details