Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020978.4 | NP_066188.1 | 3 | 744 | |
GenBank mRNA | AK095605.1 | - | 2 | 1414 | |
AK127047.1 | - | 2 | 2183 | ||
BC011179.1 | - | 3 | 436 | 4150646 | |
BC020861.1 | - | 2 | 208 | 4551745 | |
BX456665.2 | - | 3 | 688 | ||
M24895.1 | - | 1 | 118 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000699.3 | NP_000690.1 | 1 | 181 | |
GenBank mRNA | AK223463.1 | - | 1 | 96 | |
BC007060.1 | - | 1 | 136 | 3949799 | |
BC146997.1 | - | 1 | 181 | 9056925 | |
M28443.1 | - | 1 | 85 |