Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001113239.2 | NP_001106710.1 | 2 | 869 | |
NM_022740.4 | NP_073577.3 | 2 | 869 | ||
XM_006715935.2 | XP_006715998.1 | 3 | 858 | ||
XM_011516077.2 | XP_011514379.1 | 2 | 1447 | ||
XM_011516078.2 | XP_011514380.1 | 2 | 1448 | ||
XM_011516079.2 | XP_011514381.1 | 2 | 1447 | ||
XM_011516080.2 | XP_011514382.1 | 2 | 1448 | ||
XM_011516081.2 | XP_011514383.1 | 2 | 844 | ||
XM_017012069.1 | XP_016867558.1 | 2 | 950 | ||
GenBank mRNA | AF208291.1 | - | 2 | 706 | |
AF326592.1 | - | 2 | 753 |