Gene Transcripts
- Gene Symbol
- CNTNAP3
- Entrez Gene ID
- 79937
- Gene Name
- contactin associated protein-like 3
- Gene Aliases
- CASPR3, CNTNAP3A
- Chromosome Location
- Chr. 9: 39072767 - 39288303 on Build GRCh38
- UniGene ID
- Hs.128474
- Species
- Homo sapiens
- Gene Symbol
- CNTNAP3B
- Entrez Gene ID
- 728577
- Gene Name
- contactin associated protein-like 3B
- Gene Aliases
- Chromosome Location
- Chr. 9: 41891887 - 42129504 on Build GRCh38
- UniGene ID
- Hs.521495
- Species
- Homo sapiens
- Gene Symbol
- CNTNAP3P2
- Entrez Gene ID
- 643827
- Gene Name
- contactin associated protein-like 3 pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 9: 67059460 - 67297156 on Build GRCh38
- UniGene ID
- Hs.521495
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_111893.2 |
|
8 |
1283 |
|
GenBank mRNA |
AK054645.1 |
- |
8 |
1447 |
|
- Gene Symbol
- LOC100289279
- Entrez Gene ID
- 100289279
- Gene Name
- contactin associated protein-like 3-like
- Gene Aliases
- Chromosome Location
- Chr. 9: 61330592 - 61461349 on Build GRCh38
- UniGene ID
- Hs.571598
- Species
- Homo sapiens
View Full Product Details