Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005272.3 | NP_005263.1 | 5 | 781 | |
XM_011541264.2 | XP_011539566.1 | 6 | 872 | ||
GenBank mRNA | AF493909.1 | - | 5 | 568 | |
BC000233.1 | - | 5 | 749 | 3350818 | |
BT009844.1 | - | 5 | 568 |